Learn More
ATCC ZR-75-1; Breast Carcinoma; Human (Homo sapiens)
Catalog No. 502382200
Encompass
ZR-75-1 is an epithelial cell line that was isolated from the mammary gland tissue of a 63-year-old, White, female patient with ductal carcinoma. This cell line can be used in cancer research.
Catalog No. 50-238-2200 Supplier ATCC Supplier No. CRL1500
ZR-75-1 is an epithelial cell line that was isolated from the mammary gland tissue of a 63-year-old, White, female patient with ductal carcinoma. This cell line can be used in cancer research.