Learn More
Zymo Research Corporation ZR-96 DNA Sequencing Clean-up Kit™ (2 x 96 Preps)
SDP
Catalog No. 501251419
Encompass_Preferred
The ZR-96 DNA Sequencing Clean-up Kit™ employs a single-buffer system that allows for efficient DNA adsorption onto the matrix of the supplied Zymo-Spin™ IB-96 Plate. The DNA is washed then eluted with a small volume of water or loading dye containing formamide. The entire DNA purification procedure typically takes about 10 minutes.Specifications Format Filtration plate. Sample Sources Dye terminator sequencing reactions: Big Dye Terminator and other fluorescent based sequencing reactions. DNA Purity High quality, sequencing-ready DNA products that are free of unincorporated dye terminators, dNTPs, enzymes, and residual salts. Equipment Centrifuge w/ microplate carriers
Catalog No. 50-125-1419 Supplier Zymo Research Corporation Supplier No. D4052
The ZR-96 DNA Sequencing Clean-up Kit™ employs a single-buffer system that allows for efficient DNA adsorption onto the matrix of the supplied Zymo-Spin™ IB-96 Plate. The DNA is washed then eluted with a small volume of water or loading dye containing formamide. The entire DNA purification procedure typically takes about 10 minutes.Specifications Format Filtration plate. Sample Sources Dye terminator sequencing reactions: Big Dye Terminator and other fluorescent based sequencing reactions. DNA Purity High quality, sequencing-ready DNA products that are free of unincorporated dye terminators, dNTPs, enzymes, and residual salts. Equipment Centrifuge w/ microplate carriers