Learn More
Zymo Research Corporation ZR-96 Oligo Clean & Concentrator (4x96)
SDP
Catalog No. 501251421
Encompass_Preferred
The ZR-96 Oligo Clean & Concentrator™ kit features Zymo-Spin plate technology and employs a single-buffer system that allows for efficient oligonucleotide adsorption to the matrix of each well in the Zymo-Spin™I-96 Plate. There is no need for organic denaturants or chloroform. Oligonucleotide is washed and concentrated into a small volume of water ( 10 l). Purified oligonucleotide is suitable for hybridization, gel shift assays, enzymatic reactions, ligation, sequencing, microarray analysis, etc.
Catalog No. 50-125-1421 Supplier Zymo Research Corporation Supplier No. D4063
The ZR-96 Oligo Clean & Concentrator™ kit features Zymo-Spin plate technology and employs a single-buffer system that allows for efficient oligonucleotide adsorption to the matrix of each well in the Zymo-Spin™I-96 Plate. There is no need for organic denaturants or chloroform. Oligonucleotide is washed and concentrated into a small volume of water ( 10 l). Purified oligonucleotide is suitable for hybridization, gel shift assays, enzymatic reactions, ligation, sequencing, microarray analysis, etc.