Learn More
Zymo Research Corporation ZR-96 RNA Clean & Concentrator™ Kit (2 x 96 Preps)
SDP
Catalog No. 50444610
Encompass_Preferred
The kit allows for efficient RNA clean-up with the supplied Zymo-Spin™ I96 Plate. RNA is washed, then eluted and concentrated into 10 l of DNase/RNase-free Water. The highly concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.
Catalog No. 50-444-610 Supplier Zymo Research Corporation Supplier No. R1080
The kit allows for efficient RNA clean-up with the supplied Zymo-Spin™ I96 Plate. RNA is washed, then eluted and concentrated into 10 l of DNase/RNase-free Water. The highly concentrated, purified RNA is suitable for all subsequent analyses and molecular manipulations.