Learn More
Zymo Research Corporation Quick-RNA Viral 96™ Kit (4 x 96 Preps)
SDP
Supplier:  Zymo Research Corporation R1041
Encompass_Preferred
The Quick-RNA Viral 96 Kit provides for rapid, large-scale isolation of viral RNA from plasma, serum, cell culture media, cellular suspensions, urine, blood, saliva and any other biological samples stored in DNA/RNA Shield. DNA/RNA Shield ensures nucleic acid stability during sample storage/transport at ambient temperatures. The reagent effectively lyses cells and inactivates nucleases and infectious agents (virus). The kit features a specialized buffer system that facilitates complete viral particle lysis for efficient RNA isolation from samples containing enteroviruses, rhinoviruses, coronaviruses, HIV, HCV, influenza A virus, flaviviruses, measles virus, parainfluenza virus, parvovirus (a ssDNA virus), etc. Viral RNA is bound to the matrix in each well of the spin-plate, washed and eluted. The isolated high-quality viral RNA is ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/PCR detection.
Catalog No. 50-444-591