Learn More
New England Biolabs, Inc. ZraI – 1000 units
SDP
Catalog No. 501526439
Encompass
A restriction endonuclease that recognizes the sequence GAC_^GTC.
Catalog No. 50-152-6439 Supplier New England Biolabs, Inc. Supplier No. R0659L
A restriction endonuclease that recognizes the sequence GAC_^GTC.