Learn More
New England Biolabs, Inc. ZraI – 200 units
SDP
Catalog No. 50994890
Encompass
A restriction endonuclease that recognizes the sequence GAC_^GTC.
Catalog No. 50-994-890 Supplier New England Biolabs, Inc. Supplier No. R0659S
A restriction endonuclease that recognizes the sequence GAC_^GTC.