Learn More
Aviva Systems Biology Corp ZRANB2 Antibody - middle region (ARP35779_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35779_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZRANB2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZRANB2. Alternate names: DKFZp686J1831, DKFZp686N09117, ZIS, ZIS1, ZIS2, ZRANB2, ZNF265ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53020