Learn More
Aviva Systems Biology Corp ZRANB2 Antibody - middle region (ARP41219_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41219_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZRANB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZRANB2. Alternate names: DKFZp686J1831, DKFZp686N09117, FLJ41119, ZIS, ZIS1, ZIS2, ZNF265ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54059