Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zro2 Nanoparticles 5 Nm Core 50 Wt. % In Ethyl Acetate High Surface Coverage Capping Materials

Catalog No. 91600550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-600-550G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-600-550G Supplier Sigma Aldrich Supplier No. 91600550G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

High refractive index nanocrystal dispersions dramatically improve optical and mechanical properties to enable next-generation products. Incorporating ZrO2 significantly increases the refractive index of many commonly used polymers, while maintaining high transparency. This combination of key properties allows researchers to optimize optical efficiency and improve light extraction.

Specifications

CAS 1314-2-3--4
Linear Formula ZrO2
Quantity 50 g
MDL Number MFCD00011310
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.