Learn More
Sigma Aldrich Zro2 Nanoparticles 5 Nm Core Methacrylate Functionalized 50 Wt.% Propylene Glycol Methyl Ether Acetate (pgmea)
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 1314-2-3--4 |
| Linear Formula | ZrO2 |
| Quantity | 50 g |
| MDL Number | MFCD00011310 |
| Biological Activity | Respiratory system |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.