Learn More
Aviva Systems Biology Corp ZRSR2 Antibody - C-terminal region (ARP40573_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40573_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZRSR2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ZRSR2. Alternate names: URP, U2AF1L2, U2AF1RS2, U2AF1-RS2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57529