Learn More
Zymo Research Corporation Zymo-Spin™ I-96 Plate (2 Plates)
SDP
Catalog No. 50444080
Encompass_Preferred
Its deep-well, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 g of DNA, or 10 g of RNA in 10 l eluate per well.
Catalog No. 50-444-080 Supplier Zymo Research Corporation Supplier No. C2004
Its deep-well, durable polypropylene construction and unique silica-based matrix make it perfect for purifying up to 5 g of DNA, or 10 g of RNA in 10 l eluate per well.