Learn More
Zymo Research Corporation Zymo-Spin™ IIC Columns (250 Pack) (Capped)
SDP
Catalog No. 50444056
Encompass_Preferred
The Zymo-Spin IIC features durable polypropylene construction with an attached cap and contains a unique silica-based matrix that allows purification of up to 25 g of DNA or 50 g of RNA, in 25 l eluate. Capacity is 900 l.
Catalog No. 50-444-056 Supplier Zymo Research Corporation Supplier No. C1011250
The Zymo-Spin IIC features durable polypropylene construction with an attached cap and contains a unique silica-based matrix that allows purification of up to 25 g of DNA or 50 g of RNA, in 25 l eluate. Capacity is 900 l.