Learn More
Zymo Research Corporation Zymo-Spin™ IIN Columns (250 Pack) (Uncapped)
SDP
Catalog No. 50444070
Encompass_Preferred
The Zymo-Spin IIN features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 50 g of RNA, in 25 l eluate. Capacity is 900 l.
Catalog No. 50-444-070 Supplier Zymo Research Corporation Supplier No. C1019250
The Zymo-Spin IIN features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 50 g of RNA, in 25 l eluate. Capacity is 900 l.