Learn More
Zymo Research Corporation Zymo-Spin IIN Columns, 50 pack
SDP
Catalog No. 50444071
Encompass_Preferred
The Zymo-Spin IIN features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 µg of DNA, and 50 µg of RNA, in ≥ 25 µl eluate. Capacity is 900 µl.
Catalog No. 50-444-071 Supplier Zymo Research Corporation Supplier No. C101950
The Zymo-Spin IIN features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 µg of DNA, and 50 µg of RNA, in ≥ 25 µl eluate. Capacity is 900 µl.