Learn More
Zymo Research Corporation Zymo-Spin™ V Columns w/ Reservoir (25 Pack)
SDP
Catalog No. 50444064
Encompass_Preferred
The spin-column and reservoir feature durable polypropylene construction, and features a unique silica-based matrix for the purification of up to 100 g DNA or RNA in 150 l eluate. Capacity of the spin-column with reservoir is 15 ml.
Catalog No. 50-444-064 Supplier Zymo Research Corporation Supplier No. C101625
The spin-column and reservoir feature durable polypropylene construction, and features a unique silica-based matrix for the purification of up to 100 g DNA or RNA in 150 l eluate. Capacity of the spin-column with reservoir is 15 ml.