Learn More
Aviva Systems Biology Corp ZSCAN1 Antibody - middle region (ARP40057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZSCAN1. Alternate names: FLJ33779, MGC104472, MZF-1, ZNF915ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57281