Learn More
Aviva Systems Biology Corp Zscan12 Antibody - C-terminal region (ARP37222_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37222_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zscan12. It was validated on Western Blot. Zscan12,zinc finger and SCAN domain containing 12,Zfp96; FPM315; mKIAA0426; 2510038J07Rik; Zscan12,zinc finger and SCAN domain-containing protein 12,zinc finger and SCAN domain-containing protein 12,zfp-96,OTTMUSP00000000442,OTTMUSP00000000443,zinc finger protein 96The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2510038J07Rik, FPM315, Zfp96, mKIAA0426ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-600