Learn More
Aviva Systems Biology Corp Zscan12 Antibody - middle region (ARP40153_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40153_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zscan12. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2510038J07Rik, FPM315, Zfp96, mKIAA0426ApplicationWB. Species reactivity: Mouse
Catalog No. 50-284-856