Learn More
Aviva Systems Biology Corp ZSCAN16 Antibody - C-terminal region (ARP39647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZSCAN16. Alternate names: FLJ22191, ZNF392, ZNF435, dJ265C24.3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53147