Learn More
Aviva Systems Biology Corp ZSCAN2 Antibody - N-terminal region (ARP32891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZSCAN2. Alternate names: ZFP29, ZNF854ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54809