Learn More
Aviva Systems Biology Corp ZSCAN20 Antibody - middle region (ARP33522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZSCAN20. Alternate names: KOX29, ZFP-31, ZNF31, ZNF360ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55054