Learn More
Aviva Systems Biology Corp ZSCAN21 Antibody - C-terminal region (ARP39869_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39869_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZSCAN21. Alternate names: DKFZp434L134, DKFZp686H10254, NY-REN-21, ZSCAN21, Zipro1, ZNF38ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57213