Learn More
Aviva Systems Biology Corp ZSCAN5 Antibody - middle region (ARP36031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZSCAN5. Alternate names: ZNF495, ZSCAN5ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55978