Learn More
TARGETMOL CHEMICALS INC ZSTK474 200MG
SDP
Catalog No. 502579507
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PI3K Inhibitor ZSTK474 is an orally available s-triazine derivative ATP-competitive phosphatidylinositol 3-kinase (PI3K) inhibitor with potential antineoplastic activity. purity: 99%
Catalog No. 50-257-9507 Supplier TARGETMOL CHEMICALS INC Supplier No. T6168200MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PI3K Inhibitor ZSTK474 is an orally available s-triazine derivative ATP-competitive phosphatidylinositol 3-kinase (PI3K) inhibitor with potential antineoplastic activity. purity: 99%