Learn More
TARGETMOL CHEMICALS INC ZT-1A 5MG
SDP
Catalog No. 502580047
Encompass_Preferred
Also available in 1 mg and bulk. Please contact Fisher for quotes. ZT-1a is a SPAK inhibitor.ZT-1a inhibits SPAK.ZT-1a can be used for the prevention and treatment of neurodegenerative and neurocognitive disorders. purity: 99%
Catalog No. 50-258-0047 Supplier TARGETMOL CHEMICALS INC Supplier No. T395665MG
Also available in 1 mg and bulk. Please contact Fisher for quotes. ZT-1a is a SPAK inhibitor.ZT-1a inhibits SPAK.ZT-1a can be used for the prevention and treatment of neurodegenerative and neurocognitive disorders. purity: 99%