Learn More
Medchemexpress LLC Zt 52656a hydrochloride | 115730-24-0 | 99.98% | 390.87 | C19H26ClF3N2O | 10 MG
SDP
Catalog No. 5000246014
Encompass_Preferred
ZT 52656A hydrochloride is the hydrochloride salt of a selective kappa-opioid receptor agonist used in research on kappa opioid receptor pharmacology. It is reported for prevention or alleviation of ocular pain and is supplied for in vitro and in vivo experimental use.
  • Selective kappa-opioid receptor agonist, useful for KOR pharmacology studies.
  • High reported purity of 99.98% for consistent experimental results.
  • Chemical formula C19H26ClF3N2O and molecular weight 390.87 aid in dosing and calculations.
  • Available in multiple pack sizes and as a DMSO solution for flexible experimental use.
  • Provided as the hydrochloride salt to improve solubility in aqueous media.
Catalog No. 50-002-46014 Supplier Medchemexpress LLC Supplier No. HY10158210MG
ZT 52656A hydrochloride is the hydrochloride salt of a selective kappa-opioid receptor agonist used in research on kappa opioid receptor pharmacology. It is reported for prevention or alleviation of ocular pain and is supplied for in vitro and in vivo experimental use.
  • Selective kappa-opioid receptor agonist, useful for KOR pharmacology studies.
  • High reported purity of 99.98% for consistent experimental results.
  • Chemical formula C19H26ClF3N2O and molecular weight 390.87 aid in dosing and calculations.
  • Available in multiple pack sizes and as a DMSO solution for flexible experimental use.
  • Provided as the hydrochloride salt to improve solubility in aqueous media.