Learn More
Medchemexpress LLC Zt 52656a hydrochloride | 115730-24-0 | MFCD00673899 | 100.0% | 390.87 g/mol | C19H26ClF3N2O | 1 ML
SDP
Catalog No. 5000249111
Encompass_Preferred
ZT 52656A hydrochloride is a selective kappa opioid receptor agonist used in research to prevent or alleviate ocular pain. The compound is provided as a solid or as a ready-to-use 10 mM solution in DMSO, with high reported purity and manufacturer guidance for storage to maintain stability.
  • Selective kappa opioid receptor agonist suitable for ocular pain studies.
  • Available as solid or pre-dissolved 10 mM solution in DMSO for convenience.
  • High reported purity for consistent experimental results.
  • Multiple pack sizes accommodate small- and large-scale experiments.
  • Defined storage recommendations help preserve compound stability in solid and solution forms.
Catalog No. 50-002-49111 Supplier Medchemexpress LLC Supplier No. HY10158210MM 1ML
ZT 52656A hydrochloride is a selective kappa opioid receptor agonist used in research to prevent or alleviate ocular pain. The compound is provided as a solid or as a ready-to-use 10 mM solution in DMSO, with high reported purity and manufacturer guidance for storage to maintain stability.
  • Selective kappa opioid receptor agonist suitable for ocular pain studies.
  • Available as solid or pre-dissolved 10 mM solution in DMSO for convenience.
  • High reported purity for consistent experimental results.
  • Multiple pack sizes accommodate small- and large-scale experiments.
  • Defined storage recommendations help preserve compound stability in solid and solution forms.