Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zt 52656a hydrochloride | 115730-24-0 | MFCD00673899 | 100.0% | 390.87 g/mol | C19H26ClF3N2O | 25 MG
SDP

Catalog No. 5000247772
Encompass_Preferred

ZT 52656A hydrochloride is a selective kappa-opioid receptor agonist supplied as the hydrochloride salt for preclinical and in vitro research, including studies of ocular pain. The compound is for research use only and is not intended for human therapeutic use.

  • Selective kappa-opioid receptor agonist.
  • Provided as the hydrochloride salt.
  • High purity (99.98%).
  • Molecular weight 390.87 g/mol.
  • Chemical formula C19H26ClF3N2O.
  • Appearance: solid, white to off-white.
  • Available in small research sizes, e.g., 25 MG.
  • Intended for preclinical and in vitro research use only.

Catalog No. 50-002-47772 Supplier Medchemexpress LLC Supplier No. HY10158225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZT 52656A hydrochloride is a selective kappa-opioid receptor agonist supplied as the hydrochloride salt for preclinical and in vitro research, including studies of ocular pain. The compound is for research use only and is not intended for human therapeutic use.

  • Selective kappa-opioid receptor agonist.
  • Provided as the hydrochloride salt.
  • High purity (99.98%).
  • Molecular weight 390.87 g/mol.
  • Chemical formula C19H26ClF3N2O.
  • Appearance: solid, white to off-white.
  • Available in small research sizes, e.g., 25 MG.
  • Intended for preclinical and in vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.