Learn More
TARGETMOL CHEMICALS INC ZT 52656A hydrochloride 1MG
SDP
Catalog No. 502878742
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ZT 52656A is a selective agonist of kappa opioid, and used for the prevention or alleviation of pain in the eye. Purity 99.7%
Catalog No. 50-287-8742 Supplier TARGETMOL CHEMICALS INC Supplier No. T134141MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ZT 52656A is a selective agonist of kappa opioid, and used for the prevention or alleviation of pain in the eye. Purity 99.7%