Learn More
Medchemexpress LLC ZTA-261 | 3049368-51-3 | 99.9% | 428.32 | 5 MG
SDP
Catalog No. 5000391016
Encompass_Preferred
ZTA-261 is a highly selective and low toxic thyroid hormone receptor β subtype (THR-β) agonist (IC50=6.3 nM) that reduces body weight and visceral fat through a liothyronine (HY-A0070A) mediated pathway. ZTA-261 can be used in the study of lipid metabolism and obesity prevalence.
  • Highly selective and low toxic THR-β agonist.
  • Reduces body weight and visceral fat.
  • Useful in the study of lipid metabolism and obesity prevalence.
Catalog No. 50-003-91016 Supplier Medchemexpress LLC Supplier No. HY1639455MG
ZTA-261 is a highly selective and low toxic thyroid hormone receptor β subtype (THR-β) agonist (IC50=6.3 nM) that reduces body weight and visceral fat through a liothyronine (HY-A0070A) mediated pathway. ZTA-261 can be used in the study of lipid metabolism and obesity prevalence.
  • Highly selective and low toxic THR-β agonist.
  • Reduces body weight and visceral fat.
  • Useful in the study of lipid metabolism and obesity prevalence.