Learn More
Medchemexpress LLC Zta-261 | 3049368-51-3 | 99.9% | 428.32 | C21H14F6O3 | 10 MG
SDP
Catalog No. 5000289876
Encompass_Preferred
ZTA-261 is a selective thyroid hormone receptor β (THR-β) agonist used in preclinical research on lipid metabolism and obesity. It is provided as a powder for laboratory use and has been characterized for potency and composition.
  • Selective thyroid hormone receptor β (THR-β) agonist.
  • Potent activity with IC50 = 6.3 nM.
  • High chemical purity (99.9%).
  • Molecular weight 428.32 and formula C21H14F6O3.
  • Suitable for in vitro and in vivo lipid metabolism studies.
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-89876 Supplier Medchemexpress LLC Supplier No. HY16394510MG
ZTA-261 is a selective thyroid hormone receptor β (THR-β) agonist used in preclinical research on lipid metabolism and obesity. It is provided as a powder for laboratory use and has been characterized for potency and composition.
  • Selective thyroid hormone receptor β (THR-β) agonist.
  • Potent activity with IC50 = 6.3 nM.
  • High chemical purity (99.9%).
  • Molecular weight 428.32 and formula C21H14F6O3.
  • Suitable for in vitro and in vivo lipid metabolism studies.
  • Available in small research pack sizes, including 10 mg.