Learn More
Medchemexpress LLC Zuclomiphene citrate | 7619-53-6 | MFCD00082252 | 99.8% | 598.1 g/mol | C32H36ClNO8 | 10 MG
SDP
Catalog No. 5000296408
Encompass_Preferred
Zuclomiphene citrate is the cis isomer of clomiphene citrate. It is orally active and exhibits antiestrogenic activity, and is used as a research reagent in studies of estrogen receptor modulation, endocrinology, and metabolic effects such as cholesterol regulation.
  • Orally active antiestrogenic compound used in receptor modulation studies.
  • Citrate salt form, provided as a solid for laboratory research.
  • High purity: specification >95%, HPLC result 99.78% (reported).
  • Molecular formula C32H36ClNO8; molecular weight 598.1 g/mol.
  • Typically supplied in small research quantities such as 10 MG.
Catalog No. 50-002-96408 Supplier Medchemexpress LLC Supplier No. HYB1617A10MG
Zuclomiphene citrate is the cis isomer of clomiphene citrate. It is orally active and exhibits antiestrogenic activity, and is used as a research reagent in studies of estrogen receptor modulation, endocrinology, and metabolic effects such as cholesterol regulation.
  • Orally active antiestrogenic compound used in receptor modulation studies.
  • Citrate salt form, provided as a solid for laboratory research.
  • High purity: specification >95%, HPLC result 99.78% (reported).
  • Molecular formula C32H36ClNO8; molecular weight 598.1 g/mol.
  • Typically supplied in small research quantities such as 10 MG.