Learn More
Medchemexpress LLC Zuclomiphene-d4 (citrate) | 2714316-71-7 | 99.0% | 1 MG
SDP
Catalog No. 5000413063
Encompass_Preferred
Zuclomiphene-d4 citrate is a deuterium-labeled Zuclomiphene citrate. It is a cis isomer and is orally active, exhibiting an antiestrogenic effect. It can reduce cholesterol levels and is suitable for research in endocrinology and metabolic diseases.
  • Deuterium labeled compound
  • Orally active
  • Antiestrogenic effect
  • Can reduce cholesterol levels
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-13063 Supplier Medchemexpress LLC Supplier No. HYB1617AS1MG
Zuclomiphene-d4 citrate is a deuterium-labeled Zuclomiphene citrate. It is a cis isomer and is orally active, exhibiting an antiestrogenic effect. It can reduce cholesterol levels and is suitable for research in endocrinology and metabolic diseases.
  • Deuterium labeled compound
  • Orally active
  • Antiestrogenic effect
  • Can reduce cholesterol levels
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS