Learn More
Medchemexpress LLC Zuclomiphene-d4 citrate | 2714316-71-7 | 99.0% | 5 MG
SDP
Catalog No. 5000413064
Encompass_Preferred
Zuclomiphene-d4 citrate is a deuterium-labeled Zuclomiphene citrate, a cis isomer of Clomiphene citrate. It is orally active and exhibits an antiestrogenic effect, capable of reducing cholesterol levels. It is used for research in endocrinology and metabolic disease.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13064 Supplier Medchemexpress LLC Supplier No. HYB1617AS5MG
Zuclomiphene-d4 citrate is a deuterium-labeled Zuclomiphene citrate, a cis isomer of Clomiphene citrate. It is orally active and exhibits an antiestrogenic effect, capable of reducing cholesterol levels. It is used for research in endocrinology and metabolic disease.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.