Learn More
Zebra Technologies Corporation Label, Paper, 1.1875x0.8438in (30.2x21.4mm); DT, Z-Select 4000D, High Performance Coated, All-Temp Adhesive, 1in (25.4mm) core, 2710/roll, 6/box, Plain
Catalog No. NC3002866
Encompass_Preferred
Label, Paper, 1.1875x0.8438in (30.2x21.4mm); DT, Z-Select 4000D, High Performance Coated, All-Temp Adhesive, 1in (25.4mm) core, 2710/roll, 6/box, Plain
Catalog No. NC3002866 Supplier Zebra Technologies Corporation Supplier No. 10010037CS
Label, Paper, 1.1875x0.8438in (30.2x21.4mm); DT, Z-Select 4000D, High Performance Coated, All-Temp Adhesive, 1in (25.4mm) core, 2710/roll, 6/box, Plain