Learn More
Medchemexpress LLC Zunsemetinib | 1640282-42-3 | 99.99% | 513.92 | 250 MG
SDP
Catalog No. 5000348793
Encompass_Preferred
Zunsemetinib (CDD-450) is an orally active and selective p38α mitogen-activated protein kinase-activated protein kinase 2 (MK2) pathway inhibitor. It is utilized in research for immuno-inflammatory diseases and selectively blocks p38α MAPK activation of the proinflammatory kinase MK2.
  • Used for research of immuno-inflammatory diseases.
  • Selectively blocks p38α MAPK activation of the proinflammatory kinase MK2.
  • Inhibits in vitro osteoclast formation induced by RANKL.
  • Decreases IL-1β expression by promoting IL-1β mRNA degradation.
  • Reduces IL-1β secretion and promotes IL-1β mRNA instability.
  • Blocks LPS-induced TNF-α expression in vivo.
  • Increases bone density in vivo.
Catalog No. 50-003-48793 Supplier Medchemexpress LLC Supplier No. 250MGHY139553
Zunsemetinib (CDD-450) is an orally active and selective p38α mitogen-activated protein kinase-activated protein kinase 2 (MK2) pathway inhibitor. It is utilized in research for immuno-inflammatory diseases and selectively blocks p38α MAPK activation of the proinflammatory kinase MK2.
  • Used for research of immuno-inflammatory diseases.
  • Selectively blocks p38α MAPK activation of the proinflammatory kinase MK2.
  • Inhibits in vitro osteoclast formation induced by RANKL.
  • Decreases IL-1β expression by promoting IL-1β mRNA degradation.
  • Reduces IL-1β secretion and promotes IL-1β mRNA instability.
  • Blocks LPS-induced TNF-α expression in vivo.
  • Increases bone density in vivo.