Learn More
Medchemexpress LLC HY-139553 5mg , Zunsemetinib CAS:1640282-42-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1395535MG
Encompass_Preferred
HY-139553 5mg Zunsemetinib CAS: 1640282-42-3 Zunsemetinib (CDD-450) is an orally active and selective p38α mitogen-activated protein kinase-activated protein kinase 2 (MK2) pathway inhibitor. Zunsemetinib can be used for the research of immuno-inflammatory diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9253