Learn More
Aviva Systems Biology Corp ZW10 Antibody - N-terminal region (ARP58358_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58358_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZW10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZW10. Alternate names: HZW10, KNTC1AP, MGC149821ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-62291