Learn More
Agilent Technologies ZORBAX StableBond 300 C3, 4.6 x 50 mm, 5 µm
Supplier:  Agilent Technologies 860950909
Encompass
A reversed-phase C3 HPLC column with fully porous ZORBAX particles. StableBond technology ensures stability at low pH.
Catalog No. NC2676981