Learn More
Coast ZX600 Rechargeable Battery
Catalog No. 19012101
Change view
Click to view available options
Product Type:
Rechargeable Battery
Catalog No. Product Type
19-012-101 Rechargeable Battery
1 options
This item is not returnable. View return policy
Catalog No. 19-012-101 Supplier Coast Supplier No. 21135
This item is not returnable. View return policy
The integrated micro-USB and USB-C ports allow for the convenient charging of batteries inside or outside of the light source.
  • ZX600 Zithion-X™ Li-Ion ported rechargeable batteries are designed with integrated micro-USB and USB-C ports
  • Convenient battery charging either inside of, or outside of the light source
  • Includes a built-in charge level indicator that showcases the battery's charge status when charging (flashing red), changing color when fully charged (solid green)
  • Simply plug a micro-USB or USB-C cord into the battery port, connect the cord to an available power source, and charge
Specifications
Product Type Rechargeable Battery
Rechargeable Yes
Model ZX600
Material Lithium-ion
For Use With (Equipment) PM500R
Temperature (Metric) Range 20°C to 35°C (Storage)
Voltage 3.7 V