Learn More
Coast ZX955 Rechargeable Battery
Catalog No. 19012097
Change view
Click to view available options
Product Type:
Ported Battery
Catalog No. Product Type
19-012-097 Ported Battery
1 options
This item is not returnable. View return policy
Catalog No. 19-012-097 Supplier Coast Supplier No. 30368
This item is not returnable. View return policy
Batteries are designed with an integrated micro-USB to allow for convenient charging outside of the light source when necessary.
  • Zithion-X Li-Ion ported rechargeable battery is designed with an integrated micro- USB
  • Convenient charging of batteries outside of the light source when necessary
  • Batteries include a built-in charge level indicator showcasing the battery's charge status when charging (red), changing color when fully charged (green)
  • Simply plug a USB-C cord into the battery port, connect the cord to an available power source, and charge
Specifications
Product Type Ported Battery
Rechargeable Yes
Model ZX955
Material Lithium-ion
Temperature (Metric) Range 20°C to 35°C (Storage)
Voltage 3.7 V