Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zxh-1-161 | 2407654-51-5 | >98.0% | C26H24N6O3 | 10MG
SDP

Catalog No. 5000259530
Encompass_Preferred

ZXH-1-161 is a small-molecule cereblon (CRBN) modulator that selectively induces degradation of GSPT1 and is used as a research tool to study targeted protein degradation and related cellular effects. It demonstrates potent activity in relevant cell models and is supplied as a high-purity research sample suitable for in vitro assays.

  • Selective GSPT1 degradation in a CRBN-dependent manner.
  • Potent cellular activity (reported IC50 ≈ 39 nM in MM1.S cells).
  • High purity confirmed by HPLC (>98%).
  • Molecular formula C26H24N6O3; molecular weight ≈ 468.52 g·mol-1.
  • Soluble in DMSO for assay preparation.
  • Provided as a small-scale research sample for in vitro studies.

Catalog No. 50-002-59530 Supplier Medchemexpress LLC Supplier No. HY14715810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZXH-1-161 is a small-molecule cereblon (CRBN) modulator that selectively induces degradation of GSPT1 and is used as a research tool to study targeted protein degradation and related cellular effects. It demonstrates potent activity in relevant cell models and is supplied as a high-purity research sample suitable for in vitro assays.

  • Selective GSPT1 degradation in a CRBN-dependent manner.
  • Potent cellular activity (reported IC50 ≈ 39 nM in MM1.S cells).
  • High purity confirmed by HPLC (>98%).
  • Molecular formula C26H24N6O3; molecular weight ≈ 468.52 g·mol-1.
  • Soluble in DMSO for assay preparation.
  • Provided as a small-scale research sample for in vitro studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.