Learn More
Medchemexpress LLC ZXH-1-161 | 2407654-51-5 | 98.4% | 468.51 | 100 MG
SDP
Catalog No. 5000361652
Encompass_Preferred
ZXH-1-161 is a potent CRBN modulator that selectively induces the degradation of GSPT1 in a CRBN-dependent manner. It can inhibit the proliferation of multiple myeloma cells and exhibits antitumor activity.
  • Potent CRBN modulator.
  • Selectively induces degradation of GSPT1.
  • Inhibits proliferation of multiple myeloma cells.
  • Exhibits antitumor activity.
  • IC50 of 0.039 μM in MM1.S wild-type cells for antiproliferative activity.
  • Induces GSPT1 degradation in MM1.S cells at 0.01-10 μM (4 h).
  • Reduces GSPT1 levels in MM1.S cells in a dose-dependent manner.
Catalog No. 50-003-61652 Supplier Medchemexpress LLC Supplier No. HY147158100MG
ZXH-1-161 is a potent CRBN modulator that selectively induces the degradation of GSPT1 in a CRBN-dependent manner. It can inhibit the proliferation of multiple myeloma cells and exhibits antitumor activity.
  • Potent CRBN modulator.
  • Selectively induces degradation of GSPT1.
  • Inhibits proliferation of multiple myeloma cells.
  • Exhibits antitumor activity.
  • IC50 of 0.039 μM in MM1.S wild-type cells for antiproliferative activity.
  • Induces GSPT1 degradation in MM1.S cells at 0.01-10 μM (4 h).
  • Reduces GSPT1 levels in MM1.S cells in a dose-dependent manner.