Learn More
Medchemexpress LLC ZXH-1-161 | 2407654-51-5 | 98.4% | 468.51 | 5 MG
SDP
Catalog No. 5000362971
Encompass_Preferred
ZXH-1-161 is a potent CRBN modulator that selectively induces the degradation of GSPT1 in a CRBN-dependent manner. It can inhibit the proliferation of multiple myeloma cells and exhibits antitumor activity.
  • Potent CRBN modulator
  • Selectively induces degradation of GSPT1
  • Inhibits proliferation of multiple myeloma cells
  • Exhibits antitumor activity
Catalog No. 50-003-62971 Supplier Medchemexpress LLC Supplier No. HY1471585MG
ZXH-1-161 is a potent CRBN modulator that selectively induces the degradation of GSPT1 in a CRBN-dependent manner. It can inhibit the proliferation of multiple myeloma cells and exhibits antitumor activity.
  • Potent CRBN modulator
  • Selectively induces degradation of GSPT1
  • Inhibits proliferation of multiple myeloma cells
  • Exhibits antitumor activity