Learn More
TARGETMOL CHEMICALS INC ZXH-3-26 1MG
SDP
Catalog No. 502878397
Encompass_Preferred
ZXH-3-26 is a PROTAC composed of a Cereblon ligand, an E3 ubiquitin ligase, and a BRD4 ligand that can be used to study cancer. Purity 98.9%
Catalog No. 50-287-8397 Supplier TARGETMOL CHEMICALS INC Supplier No. T172971MG
ZXH-3-26 is a PROTAC composed of a Cereblon ligand, an E3 ubiquitin ligase, and a BRD4 ligand that can be used to study cancer. Purity 98.9%