Learn More
Medchemexpress LLC HY-122826 5mg Medchemexpress, ZXH-3-26 CAS:2243076-67-5 Purity:>98%
SDP
Catalog No. 501932573
Encompass_Preferred
Medchemexpress, HY-122826 5mg ZXH-3-26 CAS:2243076-67-5 ZXH-3-26 is a selective PROTAC BRD4 degrader with a DC 50/5h (DC 50/5h referring to half-maximal degradation after 5 hours of treatment) of ~ 5 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2573 Supplier Medchemexpress LLC Supplier No. HY1228265MG
Medchemexpress, HY-122826 5mg ZXH-3-26 CAS:2243076-67-5 ZXH-3-26 is a selective PROTAC BRD4 degrader with a DC 50/5h (DC 50/5h referring to half-maximal degradation after 5 hours of treatment) of ~ 5 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.