Learn More
TARGETMOL CHEMICALS INC ZY-19489 5MG
SDP
Catalog No. 502579288
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZY-19489 is an antimalarial compound that is a potential single-dose cure for Plasmodium falciparum and Plasmodium vivax malaria. It has received orphan drug designation from the FDA. purity: 98%
Catalog No. 50-257-9288 Supplier TARGETMOL CHEMICALS INC Supplier No. T676945MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZY-19489 is an antimalarial compound that is a potential single-dose cure for Plasmodium falciparum and Plasmodium vivax malaria. It has received orphan drug designation from the FDA. purity: 98%