Learn More
Medchemexpress LLC Zy-444 | 1802650-31-2 | 99.5% | 444.59 g·mol⁻1 | C26H28N4OS | 5 MG
SDP
Catalog No. 5000220985
Encompass_Preferred
ZY-444 is a small-molecule research compound reported to inhibit pyruvate carboxylase (PC) and to suppress tumor progression in preclinical studies. It is supplied as a high-purity powder for laboratory research use only; follow recommended storage conditions for powder and solutions.
  • Inhibits pyruvate carboxylase, useful for cancer metabolism research.
  • High reported purity of 99.5%.
  • Molecular weight 444.59 g·mol⁻1 and formula C26H28N4OS.
  • Available in small mg quantities and as DMSO solution formats.
  • Powder storage: -20°C for long term; solution storage: -80°C for prolonged stability.
Catalog No. 50-002-20985 Supplier Medchemexpress LLC Supplier No. HY1428705MG
ZY-444 is a small-molecule research compound reported to inhibit pyruvate carboxylase (PC) and to suppress tumor progression in preclinical studies. It is supplied as a high-purity powder for laboratory research use only; follow recommended storage conditions for powder and solutions.
  • Inhibits pyruvate carboxylase, useful for cancer metabolism research.
  • High reported purity of 99.5%.
  • Molecular weight 444.59 g·mol⁻1 and formula C26H28N4OS.
  • Available in small mg quantities and as DMSO solution formats.
  • Powder storage: -20°C for long term; solution storage: -80°C for prolonged stability.